Learn More
CELLTREAT Scientific Products 5mL Culture Tube and Dual Cap, PS, Sterile
SDP
Catalog No. NC2131757
Encompass_Preferred
Dual caps allow for either vented or sealed positions. Polystyrene tubes are optically clear to visualize cultures. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
This item is not returnable. View return policy
Catalog No. NC2131757 Supplier CELLTREAT Scientific Products Supplier No. 230434
Dual caps allow for either vented or sealed positions. Polystyrene tubes are optically clear to visualize cultures. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.