Learn More
Cell Systems Corporation Cultureboost-R is a human recombinant growth factor that also acts as a supplement in Cell Systems Complete Classic Medium (4Z0-500-R) and Cell Systems Medium Minus Serum (4Z3-500-R).
SDP
Catalog No. NC2611803
Encompass_Preferred
CultureBoost-R™ is a human recombinant growth factor that also acts as a supplement in Cell Systems Complete Classic Medium (4Z0-500-R) and Cell Systems Medium Minus Serum (4Z3-500-R). It is available in a 10mL vial in various Cell Systems media kits and "The System" as well as for purchase separately. Our high-quality formula, containing human growth factors and porcine heparin, is suitable for caring for extremely fastidious cells. CultureBoosy-R offers these benefits for your cell culture: it enhances cell growth across various Cell Systems media formulations, stabilizes cell membranes after freezing or passage, and promotes efficient cell attachment, activation, and polarization. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent Group (4Z0-800).
Catalog No. NC2611803 Supplier Cell Systems Corporation Supplier No. 4CB500R
CultureBoost-R™ is a human recombinant growth factor that also acts as a supplement in Cell Systems Complete Classic Medium (4Z0-500-R) and Cell Systems Medium Minus Serum (4Z3-500-R). It is available in a 10mL vial in various Cell Systems media kits and "The System" as well as for purchase separately. Our high-quality formula, containing human growth factors and porcine heparin, is suitable for caring for extremely fastidious cells. CultureBoosy-R offers these benefits for your cell culture: it enhances cell growth across various Cell Systems media formulations, stabilizes cell membranes after freezing or passage, and promotes efficient cell attachment, activation, and polarization. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent Group (4Z0-800).