Learn More
REPLIGEN CORPORATION Repligen CultureGard HF Perfusion Filter, 0.2 µm Pore Rating, Autoclavable, High Flow Rates, 12 Per Package
SDP
Catalog No. NC1967713
Encompass
The Repligen CultureGard HF Perfusion Filter is a sterilizing grade hollow fiber filter that enhances sterile protection during cell culture perfusion. It features a 0.2 µm DynaFibre membrane for rapid flow rates while minimizing the risk of clogging. This filter is autoclavable and designed to prevent airlocks, making it ideal for adding nutrients to bioreactors. Its hydrophilic nature ensures low hold-up volume, differentiating it from traditional filters.
Catalog No. NC1967713 Supplier REPLIGEN CORPORATION Supplier No. CU2M20512N
Add to Cart
Add to Cart
The Repligen CultureGard HF Perfusion Filter is a sterilizing grade hollow fiber filter that enhances sterile protection during cell culture perfusion. It features a 0.2 µm DynaFibre membrane for rapid flow rates while minimizing the risk of clogging. This filter is autoclavable and designed to prevent airlocks, making it ideal for adding nutrients to bioreactors. Its hydrophilic nature ensures low hold-up volume, differentiating it from traditional filters.