Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CultureOne™ Supplement (100X)

Catalog No. a3320201
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3320201 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3320201 Supplier Gibco™ Supplier No. A3320201
Only null left
Add to Cart
Edge
Add to Cart

CultureOne Supplement drives completion of neuronal differentiation, controls proliferation, and increases maturation of functional neurons. It helps achieve a higher percentage of differentiated neurons, reduced cell death, longer neurites, and greater functional responses.

CultureOne Supplement enhances the differentiation of neural stem cells (NSCs) to neurons and can be used with primary rodent neurons or psc-derived neurons. A superior alternative to anti-mitotic agents like Ara-C or mitomycin C, CultureOne Supplement not only eliminates more than 75% of contaminating neural progenitor cells with minimal cell death, it also accelerates neuronal maturation. Using CultureOne Supplement in neuronal differentiation medium for two weeks yields more mature, functional neurons, reduces cell clumping, and allows maintenance of robust cultures for over a month.

CultureOne Supplement is a chemically-defined, serum-free supplement, provided as a 100X liquid reagent that can be easily added to any conventional neuronal differentiation medium. It should be used in place of other enrichment approaches such as Ara-C, mitomycin-C, fluorouracil (5-FU), or DAPT treatment.

Improved downstream assays

Analysis of NSC-derived neurons and primary rodent neurons can be difficult due to the presence of overgrown progenitor cells in the culture. These invasive cells complicate imaging and electrophysiology experiments and can affect the physiological relevance of gene expression assays. CultureOne Supplement efficiently controls the proliferation of both neural progenitor cells and glial cells, resulting in neurons of the highest purity and accelerating maturation and function.

Non-toxic proliferation control

Traditional methods to control proliferating cell populations involve treatments with anti-mitotic agents like cytosine arabinoside (Ara-C), which can be toxic to neurons. CultureOne Supplement controls proliferation without cytotoxicity. Cultures treated with CultureOne Supplement show better neurite outgrowth, less cell clumping, and reduced cell death compared to those treated with Ara-C or mitomycin C.

Accelerated maturation of differentiated neurons

NSC-derived neurons and primary rodent neurons treated with CultureOne Supplement express mature physiological markers faster and develop longer neurites than untreated cultures. These neurons also have significantly higher numbers of voltage-gated calcium ion channels, important for neural maturity and excitability.

Extended culture duration

CultureOne Supplement enables maintenance of enriched, differentiated neurons for five weeks or longer, unlike conventional methods that result in proliferating progenitor cells, causing clumping and peeling off the plate.

Combine CultureOne Supplement with B-27 Plus Neuronal Culture System and achieve a cleaner, more relevant neuronal cell population, without needing to resort to application of Ara-C, mitomycin-C, DAPT, or any other additions to complete your neuronal cell culture medium.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -20°C) and protect from light.
Description Neural Cells, Serum-Free
Form Liquid
Product Type Microbiology Media
Product Line CultureOne
Quantity 5 mL
Shipping Condition Dry Ice
Tested For Endotoxin, Sterility
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.