Learn More
Revvity Health Sciences Inc CulturPlate 384-well, black opaque, sterile, tissue culture treated, case of 160
Catalog No. 509050489
Encompass
CulturPlate 384-well, black opaque, sterile, tissue culture treated, case of 160
Catalog No. 50-905-0489 Supplier Revvity Health Sciences Inc Supplier No. 6007668
Add to Cart
Add to Cart
CulturPlate 384-well, black opaque, sterile, tissue culture treated, case of 160