Learn More
Revvity Health Sciences Inc CulturPlate 384-well, black opaque, sterile, tissue culture treated, case of 50
Catalog No. 509051593
Encompass
CulturPlate 384-well, black opaque, sterile, tissue culture treated, case of 50
Catalog No. 50-905-1593 Supplier Revvity Health Sciences Inc Supplier No. 6007660
Add to Cart
Add to Cart
CulturPlate 384-well, black opaque, sterile, tissue culture treated, case of 50