Learn More
FUJIFILM Biosciences CultureSure 10mmol/L Y-27632
Catalog No. 5000450310
Encompass_Preferred
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It enhances cell viability and reduces cell death during passaging and cryopreservation Y-27632 inhibits contraction of the vascular smooth muscle infiltration of cancer cells and regulation of cell differentiation caused by a signal transduction system of ROCK Enhances survival rate after cryopreservation and cloning efficiency of human ES and iPS cells This product is Y-27632 prepared in water at 10 mmol/l and sterile-filtered No animal-derived matters are used in the process of manufacturing
Catalog No. 50-004-50310 Supplier FUJIFILM Biosciences Supplier No. 03924591
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It enhances cell viability and reduces cell death during passaging and cryopreservation Y-27632 inhibits contraction of the vascular smooth muscle infiltration of cancer cells and regulation of cell differentiation caused by a signal transduction system of ROCK Enhances survival rate after cryopreservation and cloning efficiency of human ES and iPS cells This product is Y-27632 prepared in water at 10 mmol/l and sterile-filtered No animal-derived matters are used in the process of manufacturing