Learn More
FUJIFILM Biosciences CultureSure 5mmol/L SB431542 D
Catalog No. 5000450271
Encompass_Preferred
This is a ready-to-use 5 mmol/L solution of SB431542 in dimethyl sulfoxide DMSO specifically formulated as animal-SB431542 inhibits ALK4 5 7 Reports on the simulation of formation of mono-layers differentiation and proliferation of embryonic stem cell-derived endothelial cells Otherwise this reagent when used together with thiazovivin and PD0325901 increases the reprogramming efficiency by 200 fold as well as accelerates reprogramming Ogawa K et al J Cell Sci 120 55 2007 Lin T et al Nat Methods 6 805 2009
Catalog No. 50-004-50271 Supplier FUJIFILM Biosciences Supplier No. 03324631
This is a ready-to-use 5 mmol/L solution of SB431542 in dimethyl sulfoxide DMSO specifically formulated as animal-SB431542 inhibits ALK4 5 7 Reports on the simulation of formation of mono-layers differentiation and proliferation of embryonic stem cell-derived endothelial cells Otherwise this reagent when used together with thiazovivin and PD0325901 increases the reprogramming efficiency by 200 fold as well as accelerates reprogramming Ogawa K et al J Cell Sci 120 55 2007 Lin T et al Nat Methods 6 805 2009