Learn More
FUJIFILM Biosciences CultureSure DMSO
Catalog No. 5000450341
Encompass_Preferred
This product is DMSO dimethyl sulfoxide for cell culture It can be used to dissolve compounds added to culture media and to prepare cryopreservation solutions
Catalog No. 50-004-50341 Supplier FUJIFILM Biosciences Supplier No. 03124051
This product is DMSO dimethyl sulfoxide for cell culture It can be used to dissolve compounds added to culture media and to prepare cryopreservation solutions