Learn More
FUJIFILM Biosciences CultureSure Y-27632
Catalog No. 5000450244
Encompass_Preferred
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It enhances cell viability and reduces cell death during passaging and cryopreservation Y-27632 is selective and potent ROCK inhibitor Y-27632 plays various roles including contraction of vascular smooth muscle cells via ROCK signaling pathways Furthermore it suppresses the cell death of human embryonic stem cells hESCs and human induced Pluripotent Stem Cells hiPSCs at cell dispersion and that it increases the survival rate of hESCs and hiPSCs during cryopreservation
Catalog No. 50-004-50244 Supplier FUJIFILM Biosciences Supplier No. 03024026
Y-27632 is reagent for cell culture applications including human ES/iPS cell culture It enhances cell viability and reduces cell death during passaging and cryopreservation Y-27632 is selective and potent ROCK inhibitor Y-27632 plays various roles including contraction of vascular smooth muscle cells via ROCK signaling pathways Furthermore it suppresses the cell death of human embryonic stem cells hESCs and human induced Pluripotent Stem Cells hiPSCs at cell dispersion and that it increases the survival rate of hESCs and hiPSCs during cryopreservation