Learn More
Research Products International Corp CultureTemp 37C Plate Warmer
SDP
Catalog No. 501367755
Encompass
Warming Plate is designed to maintain cell culture plates, dishes and reagents warm at 37°C on benchtop. Anodized aluminum plate surface maintains uniform temperature and is ideal for use with trypsin samples and cell culture applications. UV resistant housing allows use in cell culture hoods; with germicidal UV light; and withstands ethanol/isopropyl sterilization. Convenient 15 minute digital count down timer and CE approved.
Catalog No. 50-136-7755 Supplier Research Products International Corp Supplier No. 400770
Warming Plate is designed to maintain cell culture plates, dishes and reagents warm at 37°C on benchtop. Anodized aluminum plate surface maintains uniform temperature and is ideal for use with trypsin samples and cell culture applications. UV resistant housing allows use in cell culture hoods; with germicidal UV light; and withstands ethanol/isopropyl sterilization. Convenient 15 minute digital count down timer and CE approved.