Learn More
Electron Microscopy Sciences Culturewell Gasket, 2 Wells
SDP
Catalog No. 50930321
Encompass_Preferred
Reusable CultureWell gaskets from CultureWell Chambered Coverglass and CultureWell Chamber Coverglass Inserts are available bulk packaged. Ideal for forming wells on glass microscope slides or in polystyrene dishes and may be sterilized by autoclaving, UV or alcohol.
Wells: 2 Wells
Volume: 300-500 µl
Catalog No. 50-930-321 Supplier Electron Microscopy Sciences Supplier No. 704652R2
Reusable CultureWell gaskets from CultureWell Chambered Coverglass and CultureWell Chamber Coverglass Inserts are available bulk packaged. Ideal for forming wells on glass microscope slides or in polystyrene dishes and may be sterilized by autoclaving, UV or alcohol.
Wells: 2 Wells
Volume: 300-500 µl