Learn More
Electron Microscopy Sciences Culturewell Gasket, 4 Wells
SDP
Catalog No. 50930323
Encompass_Preferred
Reusable CultureWell gaskets from CultureWell Chambered Coverglass and CultureWell Chamber Coverglass Inserts are available bulk packaged. Ideal for forming wells on glass microscope slides or in polystyrene dishes and may be sterilized by autoclaving, UV or alcohol.
Wells: 4 Wells
Volume: 50-100 µl
Catalog No. 50-930-323 Supplier Electron Microscopy Sciences Supplier No. 704654R
Reusable CultureWell gaskets from CultureWell Chambered Coverglass and CultureWell Chamber Coverglass Inserts are available bulk packaged. Ideal for forming wells on glass microscope slides or in polystyrene dishes and may be sterilized by autoclaving, UV or alcohol.
Wells: 4 Wells
Volume: 50-100 µl