Learn More
FUJIFILM Biosciences CultuteSure 335-Triiodo-L-thyr
Catalog No. 5000450231
Encompass_Preferred
This is a type of thyroid hormone It has been reported to promote the differentiation of mesenchymal stem cells into adipocytes in vitro and to promote the maturation of cardiomyocytes derived from human iPS cells
Catalog No. 50-004-50231 Supplier FUJIFILM Biosciences Supplier No. 03226161
This is a type of thyroid hormone It has been reported to promote the differentiation of mesenchymal stem cells into adipocytes in vitro and to promote the maturation of cardiomyocytes derived from human iPS cells