Learn More
System Biosciences LLC Cumate Solution for the PiggyBac Transposon SystemCumate Solution, high concentration, 10,000x for use with PiggyBac = 500 µL
SDP
Catalog No. NC0266121
Encompass_Preferred
Control the cumate-inducible promoter used on SBI’s PiggyBac Vectors with this ready-to-add 10,000x inducer solution. Prepared specifically for use with SBI’s inducible PiggyBac Vectors, the Cumate Solution for the PiggyBac Transposon System is a ready-to-add 10,000x inducer solution.
Catalog No. NC0266121 Supplier System Biosciences LLC Supplier No. PBQM100A1
Add to Cart
Add to Cart
Control the cumate-inducible promoter used on SBI’s PiggyBac Vectors with this ready-to-add 10,000x inducer solution. Prepared specifically for use with SBI’s inducible PiggyBac Vectors, the Cumate Solution for the PiggyBac Transposon System is a ready-to-add 10,000x inducer solution.