Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumberland Swan 70% Isopropyl Alcohol

Catalog No. 19027026
Change view
Click to view available options
Volume (English):
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-027026 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-027026 Supplier Cumberland Supplier No. 1000033127

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be used as an effective first aid antiseptic

  • Contains 70% isopropyl alcohol
  • Made from the highest quality ingredients for national brand equivalent performance
  • Manufactured according to GMP

Specifications

Concentration 70%
Container Type Bottle
For Use With (Application) First aid antiseptic for minor cuts, scrapes and minor burns, used for rubbing and massaging, used to disinfect and sterilize objects and surfaces where Bacteria may form
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.