Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cumene Hydroperoxide

Catalog No. 2475021KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-750-21KG 1 kg
24-750-2100G 100 g
24-750-2500G 500 g
24-750-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 24-750-21KG Supplier Sigma Aldrich Supplier No. 2475021KG
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Epoxidation reagent for allylic alcohols and fatty acid esters, as an initiator for radical polymerization.

Specifications

CAS 80-15-9
Density 1.03 g/mL at 25°C
Boiling Point 100°C to 101°C (8 mmHg, lit.)
Molecular Formula C9H12O2
Refractive Index n20/D 1.5210
Linear Formula C6H5C(CH3)2OOH
MDL Number MFCD00002129
Quantity 1 kg
Synonym alpha,alpha-Dimethylbenzyl hydroperoxide
Molecular Weight (g/mol) 152.19 g/mol
Biological Activity Lungs, Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.