Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cuminaldehyde

Catalog No. 1351785G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-517-85G 5 g
13-517-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-517-85G Supplier Sigma Aldrich Supplier No. 1351785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cuminaldehyde has been used to study larvicidal and adulticidal toxicity of monoterpenes against culex pipiens
  • It has been used in evaluation of chemical composition, antimicrobial and antioxidant activities of essential oil and various extracts of eucalyptus gilii.cuminaldehyde increases the insulin secretion in streptozotocin-induced diabetic rats
  • It suppresses melanin formation in cultured murine b16-f10 melanoma cells

Specifications

CAS 122-03-2
Density 0.981 g/mL at 20 C, 0.977 g/mL at 25°C (lit.)
Boiling Point 235°C to 236°C (lit.)
Molecular Formula C10H12O
Refractive Index n20/D 1.529 (lit.)
Linear Formula (CH3)2CHC6H4CHO
MDL Number MFCD00006953
Quantity 5 g
Synonym 4-Isopropylbenzaldehyde
Molecular Weight (g/mol) 148.2
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.