Learn More
Medchemexpress LLC Cuminaldehyde (p-isopropylbenzaldehyde) | 122-03-2 | MFCD00006953 | 99.3% | 148.21 g/mol | C10H12O | 500g
SDP
Catalog No. 5000268623
Encompass_Preferred
Cuminaldehyde is the main component of Cuminum cyminum and has multiple biological activities including anti-inflammatory anti-cancer anti-diabetic anti-injury anti-neuropathy and antibacterial effects Cuminaldehyde is an inhibitor of aldose reductase (IC50 0 00085 mg/mL) -glucosidase (IC50 0 5 mg/mL) and lipoxygenase (IC50 1370 M) Cuminaldehyde also inhibits the fibrillation of -synuclein and prevents its aggregation Cuminaldehyde has potential application value in the research of neurodegenerative diseases cancer diabetes and neuropathic pain diseases[1][2][3][4][5][6][7]
Catalog No. 50-002-68623 Supplier Medchemexpress LLC Supplier No. HYY0790500G
Cuminaldehyde is the main component of Cuminum cyminum and has multiple biological activities including anti-inflammatory anti-cancer anti-diabetic anti-injury anti-neuropathy and antibacterial effects Cuminaldehyde is an inhibitor of aldose reductase (IC50 0 00085 mg/mL) -glucosidase (IC50 0 5 mg/mL) and lipoxygenase (IC50 1370 M) Cuminaldehyde also inhibits the fibrillation of -synuclein and prevents its aggregation Cuminaldehyde has potential application value in the research of neurodegenerative diseases cancer diabetes and neuropathic pain diseases[1][2][3][4][5][6][7]