Learn More
Braintree Scientific Cunningham Mouse & Neonatal Rat Adaptor
SDP
Catalog No. 501953985
Encompass
The original Cunningham™ Mouse and Neonatal Rat Adaptor fits Stoelting Lab Standard™ and most other brands of stereotaxics. It employs light, tapered Delrin ear bars, to facilitate surgery on mice and rat pups. The Cunningham™ Mouse and Neonatal Rat Adaptor features an increased overall length to accommodate larger animals, and new specialized jaw holder cuffs, providing an alternative non-invasive means of securing the animals head in the stereotaxic stage. Fully compatible with masks for gas anesthesia. **Sold for research purposes only and not for human use**
Catalog No. 50-195-3985 Supplier Braintree Scientific Supplier No. 51625
The original Cunningham™ Mouse and Neonatal Rat Adaptor fits Stoelting Lab Standard™ and most other brands of stereotaxics. It employs light, tapered Delrin ear bars, to facilitate surgery on mice and rat pups. The Cunningham™ Mouse and Neonatal Rat Adaptor features an increased overall length to accommodate larger animals, and new specialized jaw holder cuffs, providing an alternative non-invasive means of securing the animals head in the stereotaxic stage. Fully compatible with masks for gas anesthesia. **Sold for research purposes only and not for human use**