Learn More
Premier Lab Supply 40mm XRF Sample Cups – PN# SC-4340
SDP
Catalog No. NC2059820
Encompass_Preferred
40mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-4340) – Suitable for Rigaku Primus and NEX Series; Universal – OD: 40mm; ID: 32mm; Height: 25mm; Volume: 18mL – 100 per bag
Catalog No. NC2059820 Supplier Premier Lab Supply Supplier No. SC4340
40mm Double-Open Ended XRF Sample Cup with Cup, Ring & Cap (PN# SC-4340) – Suitable for Rigaku Primus and NEX Series; Universal – OD: 40mm; ID: 32mm; Height: 25mm; Volume: 18mL – 100 per bag