Learn More
Med Vet International Soufflé Cups, Paper 3/4oz, 250/Sleeve
Catalog No. 502829428
Encompass_Preferred
Soufflé Cups: Pleated portions are made of one-piece, dry-waxed construction for durability. Rims are tightly rolled for rigidity, and the base is recessed to prevent tipping.
Catalog No. 50-282-9428 Supplier Med Vet International Supplier No. NON024215H
Soufflé Cups: Pleated portions are made of one-piece, dry-waxed construction for durability. Rims are tightly rolled for rigidity, and the base is recessed to prevent tipping.