Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-440-45-160
SDP
Catalog No. NC0978850
Encompass_Preferred
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-440-45-160; 45mm Cups PN# SC-3345; Mylar film 6.0μm; 32 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.
Catalog No. NC0978850 Supplier Premier Lab Supply Supplier No. PA44045160
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-440-45-160; 45mm Cups PN# SC-3345; Mylar film 6.0μm; 32 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.