Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupferron, Thermo Scientific™

Catalog No. AA4295430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4295430 250 g
AA4295409 10 g
AA4295418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4295430 Supplier Thermo Scientific Chemicals Supplier No. 4295430

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Separation and colorimetric detection of various metals and metal oxides including Al, Cu, Fe, Ti, and vanadates
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-20-6
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00078422
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
Synonym cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
PubChem CID 2724103
IUPAC Name N-nitroso-N-phenylhydroxylamine amine
SMILES N.ON(N=O)C1=CC=CC=C1

Specifications

Melting Point 154°C (decomposition)
Odor Odorless
Quantity 250 g
UN Number UN2811
Merck Index 14,2622
Formula Weight 155.16
Physical Form Powder
Chemical Name or Material Cupferron
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H351-H315-H319-H317
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber NC4725000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.