Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK4824212 Shop All Avantor Products
Change view
Click to view available options
Poly; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-4824-212 Poly; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-4824-212 Supplier Avantor Supplier No. 482406

missing translation for 'validMainframeMsg'

Specifications

CAS 10125-13-0
Physical Form Crystalline
Quantity 2.5 kg
Molecular Formula Cl2CuH4O2
MDL Number MFCD00149674
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILES O.O.[Cl-].[Cl-].[Cu++]
IUPAC Name copper(2+) dihydrate dichloride
Molecular Weight (g/mol) 170.48
PubChem CID 61482
ChEBI CHEBI:86318
Formula Weight 170.48
Grade ACS
Packaging Poly
Chemical Name or Material Cupric Chloride, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.