Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride, Anhydr, MP Biomedicals

Catalog No. ICN205185 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205185 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205185 Supplier MP Biomedicals, Inc Supplier No. 0520518550

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7447-39-4

Specifications

Quantity 50 g
Formula Weight 134.45
Chemical Name or Material Cupric Chloride, Anhydr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.