Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride Dihydrate (Laboratory), Fisher Chemical

Catalog No. C454500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
C454-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C454-500

missing translation for 'validMainframeMsg'

Specifications

Melting Point 100°C
Color Blue Green
CAS 10125-13-0
pH 3.0 to 3.8
Physical Form Solid
Quantity 500 g
Molecular Formula Cl2CuH4O2
MDL Number MFCD00149674
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILES O.O.[Cl-].[Cl-].[Cu++]
IUPAC Name copper(2+) dihydrate dichloride
Molecular Weight (g/mol) 170.48
PubChem CID 61482
ChEBI CHEBI:86318
Formula Weight 170.48
Grade Laboratory
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Cupric Chloride Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Toxic if swallowed. Irritating to eyes, respiratory system and skin. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Corrosive to metal in aqueous
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.