Learn More
Electron Microscopy Sciences Cupric Nitrate 1% Aqueous 500 ML
SDP
Catalog No. 5031717
Encompass_Preferred
Used in Canaliculi and Lacunae Stain, Powers, Rasmussen and Clark (1951) for bones and teeth. See Powers, M.M., Rasmusen, G.L., and Clark, G.,Anat. Rec., 111:117- 6, (1951), Clark, G. (ed), Staining Procedures, 3rd edition, William & Wilkins, Baltimore, c 1973, p. 136, Bodian, D., Anat. Rec., 65:89 – 97, 1936, or Bodian, D., Anat. Rec., 69:153 – 62, 1937
Catalog No. 50-317-17 Supplier Electron Microscopy Sciences Supplier No. 2620101
Used in Canaliculi and Lacunae Stain, Powers, Rasmussen and Clark (1951) for bones and teeth. See Powers, M.M., Rasmusen, G.L., and Clark, G.,Anat. Rec., 111:117- 6, (1951), Clark, G. (ed), Staining Procedures, 3rd edition, William & Wilkins, Baltimore, c 1973, p. 136, Bodian, D., Anat. Rec., 65:89 – 97, 1936, or Bodian, D., Anat. Rec., 69:153 – 62, 1937