Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Nitrate Hydrate, MP Biomedicals

Catalog No. ICN15350180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15350180 100 g
ICN15350190 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15350180 Supplier MP Biomedicals, Inc Supplier No. 0215350180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10031-43-3
Molecular Formula CuH6N2O9
Molecular Weight (g/mol) 241.599
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

Quantity 100 g
Formula Weight 187.6 + aq.
Chemical Name or Material Cupric Nitrate Hydrate
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.