Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Oxide, Black, Powder, CP, 96%, Spectrum™ Chemical
SDP

Catalog No. 18610019 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-019 500 g Amber Glass Bottle
18-610-016 125 g Amber Glass Bottle
18-610-015 2.5 kg Amber Glass Bottle
18-610-017 12 kg Amber Glass Bottle
18-610-018 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-610-019 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1417500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1317-38-0 | 79.55 g/mol

Chemical Identifiers

CAS 1317-38-0

Specifications

CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 500 g
Molecular Weight (g/mol) 79.55
Percent Purity 96%
Grade Not Available
Packaging Amber Glass Bottle
Chemical Name or Material Cupric Oxide, Black, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.