Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Oxide Black Powder, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S031075 Shop All Aldon Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031075 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S031075 Supplier Aldon Corporation Supplier No. IS12210
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1317-38-0
Molecular Weight (g/mol) 79.54 g/mol

Specifications

Physical Form Flakes or Powder
Quantity 25 g
Percent Purity >98%
Grade Reagent
Packaging HDPE
Chemical Name or Material Cupric Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.