Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate ACS AR Fine Crystal, Macron Fine Chemicals™

Catalog No. MK4844212 Shop All Avantor Products
Change view
Click to view available options
Poly; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-4844-212 Poly; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-4844-212 Supplier Avantor Supplier No. 484406

Specifications

CAS 7758-99-8
Physical Form Fine Crystals
Quantity 2.5 kg
Molecular Formula CuH10O9S
MDL Number MFCD00149681
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper(2+) pentahydrate sulfate
Molecular Weight (g/mol) 249.68
PubChem CID 24463
ChEBI CHEBI:31440
Formula Weight 249.68
Grade ACS
Packaging Poly
Chemical Name or Material Cupric Sulfate, 5-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.