Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cupric Sulfate, Anhydrous, Powder, 98%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Spécifications
Spécifications
| CAS Max % | 1 |
| Physical Form | Powder, Needles or Crystals |
| Quantity | 50 kg |
| Molecular Weight (g/mol) | 159.6 |
| Percent Purity | 98% |
| Grade | Reagent |
| Loss on Drying | 0.01 |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Cupric Sulfate, Anhydrous, Powder |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'