Learn More
Selleck Chemical LLC Cupric sulfate-E0162-1G
SDP
Catalog No. 503138084
Encompass_Preferred
Cupric sulfate (Copper(II) sulfate) is a salt formed by treating cupric oxide with sulfuric acid Cupric sulfate is a lewis acid catalyst commonly used to promote acid-catalyzed organic transformations
Catalog No. 50-313-8084 Supplier Selleck Chemical LLC Supplier No. E01621G
Cupric sulfate (Copper(II) sulfate) is a salt formed by treating cupric oxide with sulfuric acid Cupric sulfate is a lewis acid catalyst commonly used to promote acid-catalyzed organic transformations