Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate Pentahydrate, ≥99%, (Crystalline), MP Biomedicals

Catalog No. ICN19511791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19511791 1 kg
ICN19511783 250 g
ICN19511790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19511791 Supplier MP Biomedicals, Inc Supplier No. 0219511791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range ≥99%
Formula Weight 249.68
Percent Purity ≥99%
Chemical Name or Material Cupric Sulfate Pentahydrate
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.