Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate Pentahydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCC0900125 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCC0900125 125 g
RDCC0900500 500 g
RDCC090025A 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RDCC0900125 Supplier Ricca Chemical Company Supplier No. RDCC0900125A6
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 102
Physical Form Fine Crystals or Crystalline Powder
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Packaging Natural Poly Bottle
Chemical Name or Material Cupric Sulfate Pentahydrate
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.