Learn More
Specifications
Specifications
| Boiling Point | 150°C |
| Melting Point | 110°C |
| Color | Blue |
| pH | 3.5 to 4.5 |
| Physical Form | Solid |
| Quantity | 500 g |
| UN Number | 3077 |
| Formula Weight | 249.68 |
| Grade | Technical |
| Identification | Pass Test |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. NFPA Health:2 Flammability:0 Instability:1 |
| DOTInformation | DOT Class 9, : Miscellaneous |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.