Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Pentahydrate, Granular, USP, 98.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601920 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-920 125 g Poly Bottle
18-601-921 500 g Poly Bottle
18-601-922 2.5 kg Poly Pail
18-601-923 12 kg Poly Pail
18-601-924 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-920 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CU109125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CU109, 7758-99-8, CuSO4·5H2O

Spectrum™ Chemical Cupric Sulfate, Pentahydrate, Granular, USP is used as a mineral supplement and in topical antifungal preparations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 125 g
UN Number UN3077
Percent Purity 98.5 to 100.5%
Grade USP
Loss on Drying 33.0-36.5%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.