Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate TS, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog No. 18601912 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-912 100 mL Poly Bottle
18-601-913 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-912 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C313100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C-313, 7758-98-7

Spectrum™ Chemical Cupric Sulfate TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % 0.1%, 99.9%
Grade Chemical Solution
Packaging Poly Bottle
Quantity 100 mL
Chemical Name or Material Cupric Sulfate TS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.