Learn More
Abcam CUR 61414, Hedgehog signaling pathway antagonist, 25MG
Catalog No. 502676357
Encompass_Preferred
MW 550.7 Da, Purity >98%. Novel, cell permeable Hedgehog signaling pathway antagonist (IC₅₀ = 100-200 nM). Selectively binds to smoothened (Smo) (Ki = 44 nM). Blocks the formation of and induces the regression of basaloid lesions in skin punches and shrinks UV-induced basaloid lesions in adult mouse skin. Inhibits proliferation and induces apoptosis in tumor cells without affecting non-tumor cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6357 Supplier Abcam Supplier No. AB22296425MG
MW 550.7 Da, Purity >98%. Novel, cell permeable Hedgehog signaling pathway antagonist (IC₅₀ = 100-200 nM). Selectively binds to smoothened (Smo) (Ki = 44 nM). Blocks the formation of and induces the regression of basaloid lesions in skin punches and shrinks UV-induced basaloid lesions in adult mouse skin. Inhibits proliferation and induces apoptosis in tumor cells without affecting non-tumor cells.
The product is subject to the following: Abcam Restricted Use Statement