Learn More
Medchemexpress LLC HY-113965 10mg Medchemexpress, CUR61414 CAS:334998-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11396510MG
Encompass_Preferred
Medchemexpress, HY-113965 10mg CUR61414 CAS:334998-36-6 CUR61414 is a novel, potent and cell permeable Hedgehog signaling pathway inhibitor (IC50 =100-200 nM). CUR61414 is a small-molecule aminoproline class compound and selectively binds to smoothened (Smo) with a Ki  value of 44 nM. CUR-61414 can induce apoptosis in cancer cells without affecting neighboring non-tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9273