Learn More
Medchemexpress LLC HY-N0705 5mg Medchemexpress, Curculigoside CAS:85643-19-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07055MG
Encompass_Preferred
Medchemexpress, HY-N0705 5mg Curculigoside CAS:85643-19-2 Curculigoside is the main saponin in C. orchioide, exerts significant antioxidant, anti-osteoporosis, antidepressant and neuroprotection effects. Curculigoside possesses significant anti-arthritic effects in vivo and in vitro via regulation of the JAK/STAT/NF-κB signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0305