Learn More
Cayman Chemical Curcumenol, Purity: ≥98%, CAS: 19431-84-6, 10 mg
SDP
Catalog No. 502934727
Encompass_Preferred
A sesquiterpene; active against methicillin-resistant S. aureus (MRSA) and P. aeruginosa; cytotoxic to CEM-SS lymphoblastic leukemia cells (IC50 = 11.9 µg/ml); decreases cell viability of AGS human gastric carcinoma cells (IC50 = 263.34 µM)
Catalog No. 50-293-4727 Supplier Cayman Chemical Supplier No. 2671510 MG
A sesquiterpene; active against methicillin-resistant S. aureus (MRSA) and P. aeruginosa; cytotoxic to CEM-SS lymphoblastic leukemia cells (IC50 = 11.9 µg/ml); decreases cell viability of AGS human gastric carcinoma cells (IC50 = 263.34 µM)