Learn More
Cayman Chemical Germacrone, Purity: ≥98%, CAS: 6902-91-6, 50 mg
SDP
Catalog No. 502989860
Encompass_Preferred
A sesquiterpene with diverse biological activities; inhibits PRV replication in Vero cells from 10-150 µM; enhances doxorubicin-induced cytotoxicity and apoptosis in doxorubicin-resistant MCF-7/adr cells from 50-250 µM; reduces body weight gain, visceral fat pad weight, serum insulin and plasma glucose levels, and hepatic lipid levels in a mouse model of high-fat diet-induced obesity at 5, 10, and 20 mg/kg; decreases brain MDA levels and activity of SOD and GPX, as well as reduces infarct volume in a rat model of MCAO-induced ischemia-reperfusion injury
Catalog No. 50-298-9860 Supplier Cayman Chemical Supplier No. 3011150 MG
A sesquiterpene with diverse biological activities; inhibits PRV replication in Vero cells from 10-150 µM; enhances doxorubicin-induced cytotoxicity and apoptosis in doxorubicin-resistant MCF-7/adr cells from 50-250 µM; reduces body weight gain, visceral fat pad weight, serum insulin and plasma glucose levels, and hepatic lipid levels in a mouse model of high-fat diet-induced obesity at 5, 10, and 20 mg/kg; decreases brain MDA levels and activity of SOD and GPX, as well as reduces infarct volume in a rat model of MCAO-induced ischemia-reperfusion injury