Learn More
Cayman Chemical Curcuma zedoara Curcumol 10mg
SDP
Catalog No. 502934695
Encompass_Preferred
A sesquiterpene alcohol with diverse biological activities; a PAM of GABAA receptors that potentiates GABA-induced currents in hippocampal neurons; reduces activation of proinflammatory NF-κB and profibrotic TGF-β1/SMAD signaling pathways in RAW 264.7 cells stimulated with cigarette smoke extract from 1-200 μg/ml; inhibits proliferation of and induces apoptosis in LoVo and SW480 colorectal cancer cells; reduces tumor volume in a LoVo mouse xenograft model from 20-80 mg/kg; increases the rate of wound closure in rats with streptozotocin-induced diabetes
Catalog No. 50-293-4695 Supplier Cayman Chemical Supplier No. 2684910 MG
A sesquiterpene alcohol with diverse biological activities; a PAM of GABAA receptors that potentiates GABA-induced currents in hippocampal neurons; reduces activation of proinflammatory NF-κB and profibrotic TGF-β1/SMAD signaling pathways in RAW 264.7 cells stimulated with cigarette smoke extract from 1-200 μg/ml; inhibits proliferation of and induces apoptosis in LoVo and SW480 colorectal cancer cells; reduces tumor volume in a LoVo mouse xenograft model from 20-80 mg/kg; increases the rate of wound closure in rats with streptozotocin-induced diabetes