Learn More
Cayman Chemical CurcumaZerumbon 10mg
SDP
Catalog No. 502970792
Encompass_Preferred
A natural monocyclic sesquiterpene that potently inhibits the activation of Epstein-Barr virus by phorbol esters (IC50 = 140 nM); inhibits SHH signaling and induces apoptosis in cancer cell lines
Catalog No. 50-297-0792 Supplier Cayman Chemical Supplier No. 1540010 MG
A natural monocyclic sesquiterpene that potently inhibits the activation of Epstein-Barr virus by phorbol esters (IC50 = 140 nM); inhibits SHH signaling and induces apoptosis in cancer cell lines