Learn More
Cayman Chemical Zerumbone, Purity: ≥95%, CAS: 471-05-6, 50 mg
SDP
Catalog No. 502969037
Encompass_Preferred
A natural monocyclic sesquiterpene that potently inhibits the activation of Epstein-Barr virus by phorbol esters (IC50 = 140 nM); inhibits SHH signaling and induces apoptosis in cancer cell lines
Catalog No. 50-296-9037 Supplier Cayman Chemical Supplier No. 1540050 MG
A natural monocyclic sesquiterpene that potently inhibits the activation of Epstein-Barr virus by phorbol esters (IC50 = 140 nM); inhibits SHH signaling and induces apoptosis in cancer cell lines