Learn More
Apexbio Technology LLC Curcumin(Synonyms: Diferuloylmethane, Turmeric yellow, Natural yellow 3, CI 75300), 10mM (in 1mL DMSO), CAS: 458-37-7.
SDP
Catalog No. 5000060888
Encompass_Preferred
Curcumin (CAS 458-37-7) is a natural small molecule known to inhibit tyrosinase activity with an IC50 of 47 M It modulates multiple signaling pathways involved in tumorigenesis and inflammation influencing transcription factors growth factors cytokines receptors and enzymes Clinical trials at Phase I reported no observed toxicity at doses up to 12 g/day however oral bioavailability is limited ( 1% in Phase I/II studies) Additionally curcumin demonstrates potential for Alzheimer s research as evidenced by reducing beta-amyloid (A 40 A 42) production via downregulating presenilin-1 and GSK-3 in SH-SY5Y neuroblastoma cells
Catalog No. 50-000-60888 Supplier Apexbio Technology LLC Supplier No. A33355.1
Curcumin (CAS 458-37-7) is a natural small molecule known to inhibit tyrosinase activity with an IC50 of 47 M It modulates multiple signaling pathways involved in tumorigenesis and inflammation influencing transcription factors growth factors cytokines receptors and enzymes Clinical trials at Phase I reported no observed toxicity at doses up to 12 g/day however oral bioavailability is limited ( 1% in Phase I/II studies) Additionally curcumin demonstrates potential for Alzheimer s research as evidenced by reducing beta-amyloid (A 40 A 42) production via downregulating presenilin-1 and GSK-3 in SH-SY5Y neuroblastoma cells