Learn More
Novus Biologicals™ Curcumin

Description
- Linear Formula: [HOC6H3(OCH3)CH=CHCO]2CH2
- Molecular Weight: 368.38
- Beilstein Registry Number: 2306965
- Colour Index Number: 75300
- EC Number:207-280-5
- MDL number: MFCD00008365
- PubChem ID: 24892408
Reconstitution: Reconstitute with DMSO to bring curcumin to a final concentration of 11 mg/mL.
Specifications
Specifications
| CAS | 458-37-7 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 5 g |
| Species | Human |
| Target | Curcumin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.