Learn More
Medchemexpress LLC HY-N0005 500mg , Curcumin Indian Saffron;Turmeric yellow;Natural Yellow 3;Diferuloylmethane CAS:458-37-7 Purity:98%
SDP
Catalog No. 501492416
Encompass_Preferred
Medchemexpress, HY-N0005 500mg Curcumin Indian Saffron;Turmeric yellow;Natural Yellow 3;Diferuloylmethane CAS:458-37-7 Formula:C21H20O6 Keap1-Nrf2 , Histone acetyltransferase [6] Purity:98% Curcumin is a natural phenolic compound with impressive antioxidant properties, acts as a p300/CREB-binding protein inhibitor of histone acetyltransferase activity, and also activates nuclear factor (erythroid-2 related) factor 2 (Nrf2) . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2416 Supplier Medchemexpress LLC Supplier No. HYN0005500MG
Medchemexpress, HY-N0005 500mg Curcumin Indian Saffron;Turmeric yellow;Natural Yellow 3;Diferuloylmethane CAS:458-37-7 Formula:C21H20O6 Keap1-Nrf2 , Histone acetyltransferase [6] Purity:98% Curcumin is a natural phenolic compound with impressive antioxidant properties, acts as a p300/CREB-binding protein inhibitor of histone acetyltransferase activity, and also activates nuclear factor (erythroid-2 related) factor 2 (Nrf2) . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.